Skip to Content
Merck
All Photos(1)

Key Documents

W290238

Sigma-Aldrich

(1R)-(+)-α-Pinene

97%, FG

Synonym(s):

(+)-α-Pinene, (1R,5R)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1R,5R)-2-Pinene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2902
Beilstein:
2038653
EC Number:
Council of Europe no.:
2113
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.004
NACRES:
NA.21
Organoleptic:
woody; camphoraceous; pine
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

97%

optical activity

[α]20/D +42°, neat

autoignition temp.

491 °F

refractive index

n20/D 1.465 (lit.)

bp

155-156 °C (lit.)

mp

−62 °C (lit.)

density

0.858 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

α-pinene and β-pinene

Organoleptic

woody; camphoraceous; pine

SMILES string

CC1=CC[C@@H]2C[C@H]1C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m1/s1

InChI key

GRWFGVWFFZKLTI-RKDXNWHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1R)-(+)- α-Pinene is a volatile monoterpene mainly found in the essential oil of aromatic plants.[1][2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insecticidal properties of essential plant oils against the mosquito Culex pipiens molestus (Diptera: Culicidae).
Traboulsi AF, et al.
Pest Management Science, 58(5), 491-495 (2002)
Antimicrobial agents from plants: antibacterial activity of plant volatile oils.
Dorman HJD & Deans SG
Journal of Applied Microbiology, 88(2), 308-316 (2000)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service