Skip to Content
Merck
All Photos(1)

Key Documents

W280208

Sigma-Aldrich

2-Octanone

≥98%, FG

Synonym(s):

Hexyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COCH3
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2802
Beilstein:
635843
EC Number:
Council of Europe no.:
153
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.019
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.416 (lit.)

bp

173 °C (lit.)

mp

−16 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody; earthy; herbaceous

SMILES string

CCCCCCC(C)=O

InChI

1S/C8H16O/c1-3-4-5-6-7-8(2)9/h3-7H2,1-2H3

InChI key

ZPVFWPFBNIEHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Octanone is a volatile flavor compound reported to be found in blue cheese, acerola fruit, blue crab and cray fish.

Application

  • A novel coupling technique based on thermal desorption gas chromatography with mass spectrometry and ion mobility spectrometry for breath analysis.: This study introduces a new method for analyzing breath by coupling thermal desorption gas chromatography with mass spectrometry and ion mobility spectrometry. The method enhances the detection of volatile organic compounds, including 2-Octanone, which has potential applications in medical diagnostics (Schanzmann et al., 2023).

  • A Lifetime of a Dispenser-Release Rates of Olive Fruit Fly-Associated Yeast Volatile Compounds and Their Influence on Olive Fruit Fly (Bactrocera oleae Rossi) Attraction.: The research examines the release rates of volatile compounds, including 2-Octanone, from yeast associated with olive fruit flies. These findings are significant for developing effective pest management strategies in agriculture (Veršić Bratinčević et al., 2023).

  • Identification of volatile organic compounds related to the eating quality of cooked japonica rice.: This study identifies volatile organic compounds, including 2-Octanone, that contribute to the eating quality of cooked japonica rice. The findings provide insights for the food industry to enhance rice flavor and quality (Lee et al., 2022).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components in flavour concentrates from crayfish processing waste.
Cha YJ, et al.
Journal of the Science of Food and Agriculture, 58(2), 239-248 (1992)
Volatile flavor components from boiled crayfish (Procambarus clarkii) tail meat.
Vejaphan W, et al.
Journal of Food Science, 53(6), 1666-1670 (1988)
Volatile flavor components in crayfish waste.
Tanchotikul U & Hsieh TCY
Journal of Food Science, 54(6), 1515-1520 (1989)
Cheese Flavor, gas chromatographic and mass spectral identification of neutral components of aroma faction and blue cheese.
Day EA & Anderson DF.
Journal of Agricultural and Food Chemistry, 13(1), 2-4 (1965)
J S Hosler et al.
The Journal of experimental biology, 203(Pt 18), 2797-2806 (2000-08-23)
Recent studies of olfactory blocking have revealed that binary odorant mixtures are not always processed as though they give rise to mixture-unique configural properties. When animals are conditioned to one odorant (A) and then conditioned to a mixture of that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service