Skip to Content
Merck
All Photos(1)

Documents

W269808

Sigma-Aldrich

Methyl trans-cinnamate

≥98%, stabilized, FCC, FG

Synonym(s):

trans-Cinnamic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCO2CH3
CAS Number:
Molecular Weight:
162.19
FEMA Number:
2698
Council of Europe no.:
333
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.740
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥98%

contains

alpha-tocopherol as stabilizer

bp

260-262 °C (lit.)

mp

34-38 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; fruity; strawberry

SMILES string

COC(=O)\C=C\c1ccccc1

InChI

1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+

InChI key

CCRCUPLGCSFEDV-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Special Instruction: Repeated re-heating may cause formation of gel

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vigilio Ballabeni et al.
Fitoterapia, 81(4), 289-295 (2009-10-15)
Here we investigated the anti-inflammatory properties of Ocotea quixos essential oil and of its main components, trans-cinnamaldehyde and methyl cinnamate, in in vitro and in vivo models. Ocotea essential oil and trans-cinnamaldehyde but not methyl cinnamate significantly reduced LPS-induced NO
Kathrin Fink et al.
Journal of agricultural and food chemistry, 52(10), 3065-3068 (2004-05-13)
For the authenticity assessment of (E)-methyl cinnamate from different origins, combustion/pyrolysis-isotope ratio mass spectrometry (C/P-IRMS) was used by an elemental analyzer (EA) and on-line capillary gas chromatography coupling (HRGC-C/P-IRMS). For that reason, (E)-methyl cinnamate self-prepared from synthetic, natural, and semisynthetic
Valtcho D Zheljazkov et al.
Journal of agricultural and food chemistry, 56(1), 241-245 (2007-12-13)
A field experiment was conducted to assess yield, oil content, and composition of 38 genotypes of sweet basil ( Ocimum basilicum L.). Overall, biomass yields were high and comparable to those reported in the literature. However, basil genotypes differed significantly
Bhanu Prakash et al.
International journal of food microbiology, 153(1-2), 183-191 (2011-12-06)
The investigation deals with antifungal, antiaflatoxin and antioxidant efficacy of Zanthoxylum alatum Roxb. essential oil (EO), its two major constituents and their comparison with five commonly used organic acid preservatives. The chemical profile of EO, characterized through GC and GC-MS
L P Delbressin et al.
Archives of toxicology, 49(3-4), 321-330 (1982-03-01)
The urinary mercapturic acid excretion by female rats of methyl atropate (alpha-phenyl methyl acrylate) and methyl cinnamate (beta-phenyl methyl acrylate) has been studied. On the basis of the structures of these mercapturic acids the conclusion can be drawn that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service