Skip to Content
Merck
All Photos(4)

Key Documents

W263303

Sigma-Aldrich

(R)-(+)-Limonene

greener alternative

≥93%

Synonym(s):

(R)-(+)-Limonene, (+)-p-Mentha-1,8-diene, (+)-Carvene, (R)-4-Isopropenyl-1-methyl-1-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2633
Beilstein:
2204754
EC Number:
Council of Europe no.:
491
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.045
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

4.7 (vs air)

vapor pressure

<3 mmHg ( 14.4 °C)
1 mmHg ( 20 °C)

Assay

≥93%

optical activity

[α]20/D +120°, neat

expl. lim.

6.1 %

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.473 (lit.)

bp

176-177 °C (lit.)

density

0.842 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

orange; citrus; sweet

SMILES string

CC(=C)[C@@H]1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1

InChI key

XMGQYMWWDOXHJM-JTQLQIEISA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

  • Self-assembled nanostructured aqueous dispersions as dermal delivery systems.: Investigates the potential of (R)-(+)-Limonene in pharmaceutical formulations, showing how its properties can enhance the delivery of active compounds through the skin, suggesting applications in transdermal therapeutic systems (Hoppel et al., 2015).
  • Disclaimer

    For R&D or non-EU Food use. Not for retail sale.

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

    Storage Class Code

    3 - Flammable liquids

    WGK

    WGK 2

    Flash Point(F)

    123.8 °F - closed cup

    Flash Point(C)

    51 °C - closed cup

    Personal Protective Equipment

    dust mask type N95 (US), Eyeshields, Gloves

    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Bingyin Peng et al.
    Metabolic engineering, 47, 83-93 (2018-02-23)
    Monoterpene production in Saccharomyces cerevisae requires the introduction of heterologous monoterpene synthases (MTSs). The endogenous farnesyl pyrosphosphate synthase (FPPS; Erg20p) competes with MTSs for the precursor geranyl pyrophosphate (GPP), which limits the production of monoterpenes. ERG20 is an essential gene
    Alessia Mastrodonato et al.
    Scientific reports, 8(1), 262-262 (2018-01-13)
    Exposure to extremely low-frequency electromagnetic fields (ELFEF) influences the expression of key target genes controlling adult neurogenesis and modulates hippocampus-dependent memory. Here, we assayed whether ELFEF stimulation affects olfactory memory by modulating neurogenesis in the subventricular zone (SVZ) of the
    Zeyu Lu et al.
    Nature communications, 12(1), 1051-1051 (2021-02-18)
    In metabolic engineering, loss-of-function experiments are used to understand and optimise metabolism. A conditional gene inactivation tool is required when gene deletion is lethal or detrimental to growth. Here, we exploit auxin-inducible protein degradation as a metabolic engineering approach in
    Marina Lehmann et al.
    Behavioral ecology and sociobiology, 65(2), 205-215 (2011-02-26)
    Circadian rhythmicity plays an important role for many aspects of honey bees' lives. However, the question whether it also affects learning and memory remained unanswered. To address this question, we studied the effect of circadian timing on olfactory learning and
    Johannes Niebler et al.
    Phytochemistry, 109, 66-75 (2014-12-04)
    Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service