Skip to Content
Merck
All Photos(1)

Key Documents

W245712

Sigma-Aldrich

Ethyl pyruvate

natural, FG

Synonym(s):

Ethyl 2-oxopropanoate, Ethyl acetyl formate, Ethyl pyroracemate, Pyruvic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOC2H5
CAS Number:
Molecular Weight:
116.12
FEMA Number:
2457
Beilstein:
1071466
EC Number:
Council of Europe no.:
430c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.442
NACRES:
NA.21
Organoleptic:
fruity; ethereal; rum; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.404 (lit.)

bp

144 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; ethereal; rum; sweet

SMILES string

CCOC(=O)C(C)=O

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3

InChI key

XXRCUYVCPSWGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl pyruvate is one of the volatile flavor compounds identified in roasted cocoa and beer.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl pyruvate: A new indicator of flavor stability of beer and its controlling factors.
Shimizu C, et al.
Journal of the American Chemical Society, 63(3), 135-141 (2005)
New volatile components of roasted cocoa
Kettenes DK, et al.
Journal of Agricultural and Food Chemistry, 19(2), 276-280 (1971)
Runkuan Yang et al.
Critical care (London, England), 16(1), R9-R9 (2012-01-18)
Inflammation may critically affect mechanisms of liver injury in acetaminophen (APAP) hepatotoxicity. Kupffer cells (KC) play important roles in inflammation, and KC depletion confers protection at early time points after APAP treatment but can lead to more severe injury at
Zheng-Gang Luan et al.
The Journal of surgical research, 181(1), 76-84 (2012-06-22)
Systemic inflammatory mediators have an important role in the development of acute pancreatitis. In this study, we investigated the effect of ethyl pyruvate (EP) on pancreas injury in rats with severe acute pancreatitis (SAP) and its possible mechanism. We randomly allocated
T J Percival et al.
The British journal of surgery, 99 Suppl 1, 66-74 (2012-03-28)
Extremity injury with ischaemia is the most common pattern of vascular trauma and is a challenge for surgeons who must make decisions about the timing and mechanism of limb reperfusion. In modern military conflicts, effective use of limb tourniquets and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service