W244112
Ethyl laurate
natural, ≥98%, FCC, FG
Synonym(s):
Ethyl dodecanoate, Ethyl laurate, Lauric acid ethyl ester
About This Item
Kosher
natural
Recommended Products
grade
FG
Kosher
natural
Quality Level
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
Assay
≥98%
refractive index
n20/D 1.432 (lit.)
bp
269 °C (lit.)
density
0.86 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
floral; waxy; soapy; sweet
SMILES string
CCCCCCCCCCCC(=O)OCC
InChI
1S/C14H28O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h3-13H2,1-2H3
InChI key
MMXKVMNBHPAILY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service