Skip to Content
Merck
All Photos(1)

Key Documents

W241105

Sigma-Aldrich

4-Allylanisole

≥98%, FCC

Synonym(s):

p-Allylphenyl methyl ether, p-Methoxyallylbenzene, Chavicol methyl ether, Estragole, Isoanethole, Methyl chavicol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
1099454
EC Number:
Council of Europe no.:
184
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
anise; green; spicy; herbaceous; minty; sweet
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

composition

Contains IFRA restricted 4-Allylanisole

refractive index

n20/D 1.521 (lit.)

bp

215-216 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

anise; green; spicy; herbaceous; minty; sweet

SMILES string

COc1ccc(CC=C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3,5-8H,1,4H2,2H3

InChI key

ZFMSMUAANRJZFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David Fowler et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 366(1582), 3196-3209 (2011-10-19)
This paper reports measurements of land-atmosphere fluxes of sensible and latent heat, momentum, CO(2), volatile organic compounds (VOCs), NO, NO(2), N(2)O and O(3) over a 30 m high rainforest canopy and a 12 m high oil palm plantation in the
Wasma Alhusainy et al.
Toxicological sciences : an official journal of the Society of Toxicology, 129(1), 174-187 (2012-06-01)
The alkenylbenzene estragole is a constituent of several herbs and spices. It induces hepatomas in rodents at high doses following bioactivation by cytochrome P450s and sulfotransferases (SULTs) giving rise to the ultimate carcinogenic metabolite 1'-sulfooxyestragole which forms DNA adducts. Methanolic
Yuji Ishii et al.
Chemical research in toxicology, 24(4), 532-541 (2011-03-10)
Estragole (ES) is a natural constituent of several herbs and spices that acts as a carcinogen in the livers of rodents. Given that the proximal electrophilic form of ES with a reactive carbocation is generated by cytochrome P450 and a
M De Vincenzi et al.
Fitoterapia, 71(6), 725-729 (2000-11-15)
Estragole (ES) is a natural constituent of a number of plants (e.g. tarragon, sweet basil and sweet fennel) and their essential oils have been widely used in foodstuffs as flavouring agents. Several studies with oral, i.p. or s.c. administration to
A R MacKenzie et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 366(1582), 3177-3195 (2011-10-19)
We report measurements of atmospheric composition over a tropical rainforest and over a nearby oil palm plantation in Sabah, Borneo. The primary vegetation in each of the two landscapes emits very different amounts and kinds of volatile organic compounds (VOCs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service