Skip to Content
Merck
All Photos(1)

Key Documents

V2607

Sigma-Aldrich

Vinylene carbonate

contains ≤2% BHT as stabilizer, 97%

Synonym(s):

1,3-Dioxol-2-one, VC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2O3
CAS Number:
Molecular Weight:
86.05
Beilstein:
105683
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

contains

≤2% BHT as stabilizer

refractive index

n20/D 1.421 (lit.)

bp

162 °C (lit.)

mp

19-22 °C (lit.)

density

1.355 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1OC=CO1

InChI

1S/C3H2O3/c4-3-5-1-2-6-3/h1-2H

InChI key

VAYTZRYEBVHVLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Stomach

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthieu Groh et al.
European journal of internal medicine, 26(7), 545-553 (2015-05-15)
To develop disease-specific recommendations for the diagnosis and management of eosinophilic granulomatosis with polyangiitis (Churg-Strauss syndrome) (EGPA). The EGPA Consensus Task Force experts comprised 8 pulmonologists, 6 internists, 4 rheumatologists, 3 nephrologists, 1 pathologist and 1 allergist from 5 European
Nicola Coppola et al.
Liver international : official journal of the International Association for the Study of the Liver, 35(8), 1959-1963 (2015-01-13)
A common non-synonymous polymorphism, E167K, in transmembrane six superfamily member 2 (TM6SF2) gene has been recently associated with an increased hepatic triglyceride content, dyslipidemia and liver fibrosis in NAFLD patients. We investigated possible associations between the TM6SF2 variants and liver
Stefan Hilf et al.
Journal of the American Chemical Society, 130(33), 11040-11048 (2008-07-24)
The selective placement of a functional group at the chain end of a ring-opening metathesis polymer using ruthenium carbene initiators has been a significant limitation. Here we demonstrate a highly effective and facile end-capping technique for ROMP with living ruthenium
Nicholas S Thaler et al.
Journal of attention disorders, 19(12), 1013-1021 (2012-10-18)
Recent evidence suggests that the factor structure of the Wechsler Intelligence Scale for Children-Fourth Edition (WISC-IV) is better explained by a five-factor model rather than the four-factor model in the standardization sample. The current study examined the WISC-IV's factor structure
Tor Paaske Utheim
Stem cells (Dayton, Ohio), 33(6), 1685-1695 (2015-03-20)
A number of diseases and external factors can deplete limbal stem cells, causing pain and visual loss. Ten years have passed since the first transplantation of cultured oral mucosal epithelial cells in humans, representing the first autologous cell-based therapy for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service