Key Documents
TEM-CF100HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 100 mesh, box of 50
Synonym(s):
CARBON FILM ONLY ON 100 HEX COPPER GRIDS 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 50
size
100 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 100 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)