Skip to Content
Merck
All Photos(2)

Key Documents

T89605

Sigma-Aldrich

Tropinone

99%

Synonym(s):

8-Methyl-8-azabicyclo[3.2.1]octan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO
CAS Number:
Molecular Weight:
139.19
Beilstein:
2329
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Quality Level

Assay

99%

bp

113 °C/25 mmHg (lit.)

mp

40-44 °C (lit.)

storage temp.

2-8°C

SMILES string

CN1[C@@H]2CC[C@H]1CC(=O)C2

InChI

1S/C8H13NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-7H,2-5H2,1H3/t6-,7+

InChI key

QQXLDOJGLXJCSE-KNVOCYPGSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsuko Yamashita et al.
Biochemistry, 42(19), 5566-5573 (2003-05-14)
To understand the catalytic mechanism of an enzyme, it is crucial to determine the crystallographic structures corresponding to the individual reaction steps. Here, we report two crystal structures of enzyme-substrate complexes prior to reaction initiation: tropinone reductase-II (TR-II)-NADPH and TR-II-NADPH-tropinone
Enantioselective synthesis of unnatural (S)-(+)-cocaine.
J C Lee et al.
The Journal of organic chemistry, 65(15), 4773-4775 (2000-08-26)
E Leete
Planta medica, 56(4), 339-352 (1990-08-01)
Recent work on the biosynthesis of the tropane moiety of cocaine, hyoscamine, scopolamine, and related alkaloids is reviewed. Revision of the generally accepted biosynthetic pathway to these alkaloids is now proposed in the light of new discoveries. New information on
Franklin A Davis et al.
Organic letters, 11(7), 1647-1650 (2009-03-13)
Sulfinimine-derived, enantiopure N-sulfinyl beta-amino ketone ketals on hydrolysis give dehydropyrrolidine ketones that on treatment with (Boc)(2)O/DMAP afford substituted tropinones in good yield.
A Portsteffen et al.
Phytochemistry, 37(2), 391-400 (1994-09-01)
In tropane-alkaloid producing plants and root cultures, the reduction of tropinone is a branch-point in secondary metabolism. Two different reductases stereospecifically form the isomeric alcohols tropine (tropan-3 alpha-ol) and pseudotropine (tropan-3 beta-ol). We describe here the purification and characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service