Skip to Content
Merck
All Photos(2)

Key Documents

T30201

Sigma-Aldrich

3,3′-Thiodipropionic acid

97%

Synonym(s):

Bis(2-carboxyethyl) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(CH2CH2COOH)2
CAS Number:
Molecular Weight:
178.21
Beilstein:
1210299
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

131-134 °C (lit.)

SMILES string

OC(=O)CCSCCC(O)=O

InChI

1S/C6H10O4S/c7-5(8)1-3-11-4-2-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

ODJQKYXPKWQWNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Thiodipropionic acid is an organic disulfide that can be used as a ligand for the synthesis of cadminum and zinc coordination polymers with luminescent properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Structure, and Luminescence Properties of Zinc (II) and Cadmium (II) Complexes containing the Flexible Ligand of 3, 3??Thiodipropionic Acid.
Yang P, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 634(6?7), 1221-1224 (2008)
Hong-Seok Kim et al.
Foodborne pathogens and disease, 14(2), 84-88 (2017-01-05)
Culture-based detection of nontyphoidal Salmonella spp. in foods requires at least four working days; therefore, new detection methods that shorten the test time are needed. In this study, we developed a novel single-step Salmonella enrichment broth, SSE-1, and compared its
L C McDonald et al.
Applied and environmental microbiology, 45(2), 360-365 (1983-02-01)
Escherichia coli LSUFS was injured either by freezing at -10 degrees C or by heating at 57 degrees C for 12 min. Surviving cells were recovered on nonselective tryptone-glucose extract agar and selective violet red bile agar supplemented with compounds
Nadine Bruland et al.
The Journal of biological chemistry, 284(1), 660-672 (2008-11-13)
The thioether 3,3-thiodipropionic acid can be used as precursor substrate for biotechnological synthesis of 3-mercaptopropionic acid-containing polythioesters. Therefore, the hitherto unknown catabolism of this compound was elucidated to engineer novel and improved polythioester biosynthesis pathways in the future. Bacteria capable
Yasutaka Kamei et al.
Macromolecular bioscience, 7(3), 364-372 (2007-03-21)
To prepare sulfur-containing natural polymers effectively, several plant oils and 3,3'-thiodipropionic acid (TDP) have been used as carbon sources for the biosynthesis of copolymer poly[(3-hydroxybutyrate)-co-(3-mercaptopropionate)] [poly(3HB-co-3MP)] by a wild-type bacterium Cupriviadus necator H16. By using the plant oils, copolymer accumulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service