SYNC362640C
Synthware™ chromatography column with PTFE stopcock and coarse frit
L 457 mm
Synonym(s):
Synthware C362640C, glass chromatograpy column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C362640C
L
457 mm
Looking for similar products? Visit Product Comparison Guide
General description
With reinforced beaded top, coarse porosity fritted disc to support column packing. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service