S4808
cis-Stilbene
96%
Synonym(s):
cis-1,2-Diphenylethylene, Isostilbene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH=CHC6H5
CAS Number:
Molecular Weight:
180.25
Beilstein:
1616739
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
refractive index
n20/D 1.622 (lit.)
bp
82-84 °C/0.4 mmHg (lit.)
density
1.011 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
c1ccc(cc1)\C=C/c2ccccc2
InChI
1S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12H/b12-11-
InChI key
PJANXHGTPQOBST-QXMHVHEDSA-N
Gene Information
human ... ESR1(2099)
rat ... Esr1(24890)
Looking for similar products? Visit Product Comparison Guide
General description
cis-Stilbene can undergo epoxidation in the presence of supported gold nanoparticles (AuNP) and using cumene as a solvent to form a mixture of cis- and trans-stilbene oxides.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daniel T Bregante et al.
Journal of the American Chemical Society, 139(20), 6888-6898 (2017-04-30)
Group IV and V framework-substituted zeolites have been used for olefin epoxidation reactions for decades, yet the underlying properties that determine the selectivities and turnover rates of these catalysts have not yet been elucidated. Here, a combination of kinetic, thermodynamic
Amani A Mahbub et al.
Anti-cancer agents in medicinal chemistry, 13(10), 1601-1613 (2013-06-26)
Mortality rates for leukemia are high despite considerable improvements in treatment. Since polyphenols exert pro-apoptotic effects in solid tumors, our study investigated the effects of polyphenols in haematological malignancies. The effect of eight polyphenols (quercetin, chrysin, apigenin, emodin, aloe-emodin, rhein
Epoxidation of stilbene using supported gold nanoparticles: cumyl peroxyl radical activation at the gold nanoparticle surface.
Crites COL, et al.
Chemical Communications (Cambridge, England), 50(18), 2289-2291 (2014)
Jana L van den Berg et al.
The journal of physical chemistry. A, 124(29), 5999-6008 (2020-06-26)
Previously, it has been demonstrated that external electric fields may be used to exert control over chemical reactivity. In this study, the impact of a strong, nonresonant IR field (1064 nm) on the photoisomerization of cis-stilbene is investigated in cyclohexane
Hassan E Eldesouky et al.
Scientific reports, 10(1), 6089-6089 (2020-04-10)
Azole antifungals are vital therapeutic options for treating invasive mycotic infections. However, the emergence of azole-resistant isolates combined with limited therapeutic options presents a growing challenge in medical mycology. To address this issue, we utilized microdilution checkerboard assays to evaluate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service