Skip to Content
Merck
All Photos(1)

Key Documents

M34006

Sigma-Aldrich

3-Methylcatechol

98%

Synonym(s):

1,2-Dihydroxy-3-methylbenzene, 2,3-Dihydroxytoluene, 2,3-Toluenediol, 2-Hydroxy-3-methylphenol, 2-Hydroxy-6-methylphenol, 3-Methyl-1,2-benzenediol, 3-Methyl-1,2-dihydroxybenzene, 3-Methylpyrocatechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,2-(OH)2
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

241 °C (lit.)

mp

65-68 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cc1cccc(O)c1O

InChI

1S/C7H8O2/c1-5-3-2-4-6(8)7(5)9/h2-4,8-9H,1H3

InChI key

PGSWEKYNAOWQDF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Cerdan et al.
Journal of bacteriology, 176(19), 6074-6081 (1994-10-01)
Catechol 2,3-dioxygenase encoded by TOL plasmid pWW0 of Pseudomonas putida consists of four identical subunits, each containing one ferrous ion. The enzyme catalyzes ring cleavage of catechol, 3-methylcatechol, and 4-methylcatechol but shows only weak activity toward 4-ethylcatechol. Two mutants of
L E Hüsken et al.
Journal of biotechnology, 88(1), 11-19 (2001-05-30)
Pseudomonas putida MC2 is a solvent-tolerant strain that accumulates 3-methylcatechol. In aqueous media, 10 mM of 3-methylcatechol was produced and production was limited by 3-methylcatechol toxicity to the biocatalyst. Production levels increased by introduction of a second, organic phase that
J Wery et al.
Applied microbiology and biotechnology, 54(2), 180-185 (2000-09-01)
The aim of the study was to investigate whether toxic fine chemical production can be improved using the solvent-tolerant Pseudomonas putida S12 in a two-liquid-phase system consisting of aqueous media and a water-immiscible octanol phase with production of 3-methylcatechol from
Leonie E Hüsken et al.
Journal of biotechnology, 96(3), 281-289 (2002-06-05)
Bioproduction of 3-methylcatechol from toluene by Pseudomonas putida MC2 was studied in the presence of an additional 1-octanol phase. This solvent was used to supply the substrate and extract the product, in order to keep the aqueous concentrations low. A
M G Wallis et al.
The Biochemical journal, 266(2), 605-609 (1990-03-01)
A purification procedure has been developed for an extradiol dioxygenase expressed in Escherichia coli, which was originally derived from a Pseudomonas putida strain able to grow on toluidine. Physical and kinetic properties of the enzyme have been investigated. The enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service