All Photos(1)
About This Item
Linear Formula:
CH3OC6H4CONH2
CAS Number:
Molecular Weight:
151.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
164-167 °C (lit.)
SMILES string
COc1ccc(cc1)C(N)=O
InChI
1S/C8H9NO2/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H2,9,10)
InChI key
GUCPYIYFQVTFSI-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prashant Kesharwani et al.
Mini reviews in medicinal chemistry, 17(18), 1725-1745 (2016-02-20)
Nanoparticles (NPs), the submicron-sized colloidal particles, have recently generated enormous interest among biomedical scientists, particularly in cancer therapy. A number of models are being used for exploring NPs safety and efficacy. Recently, cancer cell lines have explored as prominent experimental
Andrew B Satterlee et al.
Theranostics, 7(2), 253-269 (2017-01-04)
Aggressive, desmoplastic tumors are notoriously difficult to treat because of their extensive stroma, high interstitial pressure, and resistant tumor microenvironment. We have developed a combination therapy that can significantly slow the growth of large, stroma-rich tumors by causing massive apoptosis
Chong Su et al.
Advanced drug delivery reviews, 143, 97-114 (2019-07-01)
Nanocarriers (NCs) are a type of drug delivery system commonly used to regulate the pharmacokinetic and pharmacodynamic properties of drugs. Although a wide variety of NCs has been developed, relatively few have been registered for clinical trials and even fewer
Hongda Zhu et al.
Nanoscale, 12(5), 3400-3410 (2020-01-29)
Highly fibrotic and collagen-rich properties in desmoplastic melanoma (DM) result in an immune-suppressive fibrotic tumor microenvironment (TME) that resists clinical therapies. The different clinical and pathological properties, as compared to conventional melanoma, lead to delayed diagnosis and it is difficult
Qinglei Meng et al.
Nature communications, 8, 14190-14190 (2017-02-01)
Cyclohexanone and its derivatives are very important chemicals, which are currently produced mainly by oxidation of cyclohexane or alkylcyclohexane, hydrogenation of phenols, and alkylation of cyclohexanone. Here we report that bromide salt-modified Pd/C in H
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service