Skip to Content
Merck
All Photos(2)

Key Documents

H48107

Sigma-Aldrich

5-Hydroxy-2-nitrobenzaldehyde

98%

Synonym(s):

2-Nitro-5-hydroxybenzaldehyde, 3-Formyl-4-nitrophenol, 3-Hydroxy-6-nitrobenzaldehyde, 5-Hydroxy-2-nitrobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)CHO
CAS Number:
Molecular Weight:
167.12
Beilstein:
1910196
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

165-169 °C (lit.)

SMILES string

[H]C(=O)c1cc(O)ccc1[N+]([O-])=O

InChI

1S/C7H5NO4/c9-4-5-3-6(10)1-2-7(5)8(11)12/h1-4,10H

InChI key

XLYPHUGUKGMURE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service