GF92536993
Gadolinium
wire, 1.0 mm diameter, purity 99.9%, length 25 mm
Synonym(s):
GD005110
About This Item
Assay
99.9%
form
wire
manufacturer/tradename
Goodfellow 925-369-93
resistivity
126 μΩ-cm, 20°C
L
25 mm
diameter
1.0 mm
bp
3273 °C (lit.)
mp
1313 °C (lit.)
density
7.886 g/mL at 25 °C (lit.)
SMILES string
[Gd]
InChI
1S/Gd
InChI key
UIWYJDYFSGRHKR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 3
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service