Skip to Content
Merck
All Photos(1)

Key Documents

GF90142769

Polyimide

(PI), sheet, thickness 1.5 mm, size 296 × 296 mm, grade cirlex cl

Synonym(s):

IM303120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

sheet

manufacturer/tradename

Goodfellow 901-427-69

size

296 × 296 mm

thickness

1.5 mm

General description

For updated SDS information please visit www.goodfellow.com
Polyimides are an important class of high-temperature engineering polymers that consist of aromatic dianhydrides and diamine monomers. These polymers are well known for their high-temperature stability, mechanical strength, chemical resistance, and excellent dielectric properties. These characteristics of the polyimides make them an ideal candidate for aerospace, flexible printed circuits, electrical insulation, high-temperature mechanical seals, photovoltaic modules, high-density electronic interconnects, and many other electronic applications. Polyimides are also used in medical tubing applications because of their high flexibility, high tensile strength, biocompatibility, low friction, transparency, smooth surface, and they can be sterilized via gamma, EtO, and e-beam sterilization methods.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyimides
The Biochemistry of the Nucleic Acids (2012)
Polyimides
F.I.L./I.D.F., Milchwiss. (2017)
Achievements in low-pressure membrane processes microfiltration (MF) and ultrafiltration (UF) for wastewater and water treatment
Current Topics in the Chemistry of Boron, 67-107 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service