Skip to Content
Merck
All Photos(2)

Key Documents

GF85947519

Neodymium

rod, 100mm, diameter 12.5mm, cast, 99%

Synonym(s):

Neodymium, ND007940

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24
MDL number:
UNSPSC Code:
12141609
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

rod

manufacturer/tradename

Goodfellow 859-475-19

resistivity

64.0 μΩ-cm, 20°C

bp

3074 °C (lit.)

mp

1021 °C (lit.)

density

7.003 g/mL at 25 °C (lit.)

SMILES string

[Nd]

InChI

1S/Nd

InChI key

QEFYFXOXNSNQGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franz Fankhauser et al.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde, 216(6), 381-398 (2003-02-05)
The physical and clinical features of a number of laser energy sources presently being used in plastic and endonasal surgery, such as the Nd:YAG, KTP, Ho:YAG, Er:YAG and CO(2) lasers, are emphasized. Particular attention is paid to the Nd:YAG laser
A J Costello et al.
European urology, 35(2), 138-146 (1999-02-06)
Not until 1991 did laser therapy for benign prostatic hyperplasia become a feasible option for the practicing urologist. Prior to the development of side firing free beam laser delivery devices contact Neodymium:YAG, laser therapy for BPH was of limited applicability.
Francisco J Muñoz-Negrete et al.
BioMed research international, 2015, 742792-742792 (2015-11-12)
Uveitic glaucoma (UG) typically is associated with very high intraocular pressure (IOP) and more intense optic nerve damage than other glaucoma types. This secondary glaucoma requires an early diagnosis and adequate management of both uveitis and glaucoma. It is mandatory
B S Stein
Mayo Clinic proceedings, 73(8), 787-791 (1998-08-14)
Since 1986, benign prostatic hyperplasia has been treated with lasers, but clinical use was not practical until the right-angled fiber was developed in the early 1990s. The neodymium:yttrium-aluminum-garnet (Nd:YAG) laser is one of four types available for treating the prostate.
J B Dickson et al.
Ophthalmic plastic and reconstructive surgery, 5(1), 17-26 (1989-01-01)
Contact Nd:YAG laser surgery offers a new method of delivering laser energy to a target tissue site. Nd:YAG, a near-infrared laser, can be transmitted through an optical fiber delivery system to a synthetic sapphire probe. This sapphire probe can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service