Skip to Content
Merck
All Photos(2)

Key Documents

GF85328523

Ruthenium

Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%

Synonym(s):

Ruthenium, RU006100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07
MDL number:
UNSPSC Code:
12141739
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 853-285-23

resistivity

7.1 μΩ-cm, 0°C

particle size

10 mm

bp

3900 °C (lit.)

mp

2310 °C (lit.)

density

12.45 g/cm3 (lit.)

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bergamo et al.
Journal of inorganic biochemistry, 106(1), 90-99 (2011-11-25)
The study of metal complexes for the treatment of cancer diseases has resulted in the identification of some unique properties of ruthenium-based compounds. Among these inorganic-based agents, two of them, namely the ruthenium(III) drugs NAMI-A and KP1019 have undertaken with
Aviva Levina et al.
Metallomics : integrated biometal science, 1(6), 458-470 (2009-11-01)
Interest in Ru anticancer drugs has been growing rapidly since NAMI-A ((ImH(+))[Ru(III)Cl(4)(Im)(S-dmso)], where Im = imidazole and S-dmso = S-bound dimethylsulfoxide) or KP1019 ((IndH(+))[Ru(III)Cl(4)(Ind)(2)], where Ind = indazole) have successfully completed phase I clinical trials and an array of other
Martin R Gill et al.
Chemical Society reviews, 41(8), 3179-3192 (2012-02-09)
In the last few decades, coordination complexes based on d(6) metal centres and polypyridyl ligand architectures been developed as structure- and site-specific reversible DNA binding agents. Due to their attractive photophysical properties, much of this research has focused on complexes
E Tfouni et al.
Current medicinal chemistry, 17(31), 3643-3657 (2010-09-18)
The discovery of the involvement of nitric oxide (NO) in several physiological and pathophysiological processes launched a spectacular increase in studies in areas such as chemistry, biochemistry, and pharmacology. As a consequence, the development of NO donors or scavengers for
Nicole L Fry et al.
Accounts of chemical research, 44(4), 289-298 (2011-03-03)
Nitric oxide (NO) can induce apoptosis (programmed cell death) at micromolar or higher doses. Although cell death via NO-induced apoptosis has been studied quite extensively, the targeted delivery of such doses of NO to infected or malignant tissues has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service