GF59830139
Rhenium
wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Synonym(s):
Rhenium, RE005106
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99.97%
form
wire
manufacturer/tradename
Goodfellow 598-301-39
L × diam.
0.2 m × 0.10 mm
bp
5596 °C (lit.)
5627 °C (lit.)
mp
3180 °C (lit.)
density
21.02 g/cm3 (lit.)
SMILES string
[ReH]
InChI
1S/Re
InChI key
WUAPFZMCVAUBPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Radiolabeling of monoclonal antibodies and fragments with technetium and rhenium.
Bioconjugate chemistry, 3(2), 91-99 (1992-03-01)
Rheumatology (Oxford, England), 46(10), 1531-1537 (2007-06-29)
Hypertrophy and inflammation of the synovium with various underlying pathologies - such as rheumatoid arthritis, osteoarthritis, haemophilia and spondyloarthropathy - can be treated successfully by radiosynoviorthesis (RSO). For medium-sized joints (shoulder, elbow, wrist, hip and ankle), the radionuclide of choice
Steroids, 72(3), 247-260 (2007-02-07)
OPLS all atom force field parameters were developed in order to model a diverse set of novel rhenium based estrogen receptor ligands whose relative binding affinities (RBA) to the estrogen receptor alpha isoform (ERalpha) with respect to 17beta-estradiol were available.
Mini reviews in medicinal chemistry, 4(1), 61-68 (2004-02-03)
The favourable nuclear properties of copper-67 and rhenium-188 for therapeutic application are described, together with methods for the chemical synthesis of a number of derivatives. Survival from invasive bladder cancer has changed little over the past 20 years. The intravesicular
Chemical communications (Cambridge, England), (5)(5), 493-512 (2009-03-14)
Radiolabeled biomolecules can be used to visualize a variety of diseases through interaction with specific cell receptors. A key step is the introduction of a molecular entity that allows facile labeling with the medically useful radionuclide (99m)Tc without significant alteration
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service