GF38737929
Tantalum
pellets, 20g, max. size 10mm, 99.9%
Synonym(s):
Tantalum, TA006100, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.9%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 387-379-29
resistivity
13.5 μΩ-cm, 20°C
max. part. size
10 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
The Journal of arthroplasty, 29(4), 831-835 (2013-10-09)
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf
Clinical use of porous tantalum in complex primary total knee arthroplasty.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Zhongguo gu shang = China journal of orthopaedics and traumatology, 26(7), 617-620 (2013-10-19)
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service