GF38519376
Holmium
foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Synonym(s):
Holmium, HO000205
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99%
form
foil
manufacturer/tradename
Goodfellow 385-193-76
size × thickness
100x100 mm × 0.0125 mm
density
8.8 g/mL at 25 °C (lit.)
SMILES string
[Ho]
InChI
1S/Ho
InChI key
KJZYNXUDTRRSPN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Urology, 76(6), 1334-1337 (2010-06-25)
To assess the outcome of flexible ureterorenoscopy (F-URS) with the holmium laser in treating stones in the horseshoe kidney (HSK). We retrospectively reviewed the records of 17 patients with a HSK stone (17 renal units) who had undergone F-URS with
A systematic review of the clinical efficacy and effectiveness of the holmium:YAG laser in urology.
BJU international, 84(1), 1-9 (1999-08-12)
World journal of urology, 25(3), 235-239 (2007-03-07)
The efficiency and safety profile of the holmium laser has made this tool a versatile multi-purpose instrument for use in the endoscopic treatment of a wide variety of urologic disorders, in particular urinary calculi. Herein we review holmium laser physics
Journal of endourology, 13(7), 477-480 (1999-11-24)
Holmium:YAG lithotripsy is effective for all stone compositions, and high success rates may be expected. Large renal and bladder calculi may be treated effectively with Ho:YAG lasertripsy. Using angled optical fibers and increasing power settings may be particularly useful to
Gastrointestinal endoscopy, 81(4), 932-942 (2014-12-17)
Cholangioscopy is used to diagnose and treat various biliary lesions. Balloon-assisted cholangioscopy (BAC) has mostly been reported in Asian patients with large bile ducts. To assess the feasibility and accuracy of performing BAC in complex biliary diseases in Australian patients.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service