GF26290313
Platinum
tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Synonym(s):
Platinum, PT007250
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.95%
form
tubes
manufacturer/tradename
Goodfellow 262-903-13
resistivity
10.6 μΩ-cm, 20°C
L × wall thickness
50 m × 0.2 mm
O.D. × I.D.
2.0 mm × 1.6 mm
bp
3827 °C (lit.)
mp
1772 °C (lit.)
density
21.45 g/cm3 (lit.)
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Platinum-free catalysts as counter electrodes in dye-sensitized solar cells.
Mingxing Wu et al.
ChemSusChem, 5(8), 1343-1357 (2012-08-14)
A G Quiroga
Journal of inorganic biochemistry, 114, 106-112 (2012-07-24)
This focused review is devoted to our advances in developing trans platinum complexes as antitumoral complexes, in particular those bearing aliphatic amines. It includes some fundamental aspects of the understanding of non conventional metallodrug mechanism. The perspective is arranged by
Xiaoyong Wang et al.
Chemical Society reviews, 42(1), 202-224 (2012-10-09)
Platinum-based anticancer drugs occupy a crucial role in the treatment of various malignant tumours. However, the efficacy and applicability of platinum drugs are heavily restricted by severe systemic toxicities and drug resistance. Different drug targeting and delivery (DTD) strategies have
Xiao-yong Shen et al.
PloS one, 8(8), e69553-e69553 (2013-08-14)
X-ray repair cross-complementing protein 3 (XRCC3) is an essential gene involved in the double-strand break repair pathway. Published evidence has shown controversial results about the relationship between XRCC3 Thr241Met polymorphism and clinical outcomes of non-small cell lung cancer (NSCLC) patients
Josep Ramon Germà-Lluch et al.
Archivos espanoles de urologia, 66(5), 524-535 (2013-06-26)
Cisplatin has been the cornerstone of germ cell testicular tumors therapy since its introduction more tan 30 years ago, and a basic part of the schemes given to multiple ovarian, lung, head and neck, and bladder tumors among others. Some
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service