Skip to Content
Merck
All Photos(2)

Key Documents

GF13376785

Holmium

rod, 200mm, diameter 3.8mm, 99%

Synonym(s):

Holmium, HO007905

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12141606
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 133-767-85

L × diam.

200 mm × 3.8 mm

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit Molimard et al.
Urology, 76(6), 1334-1337 (2010-06-25)
To assess the outcome of flexible ureterorenoscopy (F-URS) with the holmium laser in treating stones in the horseshoe kidney (HSK). We retrospectively reviewed the records of 17 patients with a HSK stone (17 renal units) who had undergone F-URS with
A systematic review of the clinical efficacy and effectiveness of the holmium:YAG laser in urology.
I Larizgoitia et al.
BJU international, 84(1), 1-9 (1999-08-12)
Sean Pierre et al.
World journal of urology, 25(3), 235-239 (2007-03-07)
The efficiency and safety profile of the holmium laser has made this tool a versatile multi-purpose instrument for use in the endoscopic treatment of a wide variety of urologic disorders, in particular urinary calculi. Herein we review holmium laser physics
J M Teichman
Journal of endourology, 13(7), 477-480 (1999-11-24)
Holmium:YAG lithotripsy is effective for all stone compositions, and high success rates may be expected. Large renal and bladder calculi may be treated effectively with Ho:YAG lasertripsy. Using angled optical fibers and increasing power settings may be particularly useful to
Peter Lim et al.
Gastrointestinal endoscopy, 81(4), 932-942 (2014-12-17)
Cholangioscopy is used to diagnose and treat various biliary lesions. Balloon-assisted cholangioscopy (BAC) has mostly been reported in Asian patients with large bile ducts. To assess the feasibility and accuracy of performing BAC in complex biliary diseases in Australian patients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service