Skip to Content
Merck
All Photos(2)

Key Documents

GF09165786

Lanthanum

foil, 25x25mm, thickness 0.1mm, as rolled, 99%

Synonym(s):

Lanthanum, LA000210

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
MDL number:
UNSPSC Code:
12141607
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99%

form

foil

manufacturer/tradename

Goodfellow 091-657-86

resistivity

54 μΩ-cm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tilman B Drüeke
Seminars in dialysis, 20(4), 329-332 (2007-07-20)
Controlling serum phosphorus levels in patients with renal failure is critical. The use of oral phosphate-binding agents is universal for patients with end-stage kidney disease to reduce phosphate absorption. The therapeutic goal is to reduce serum phosphorus levels without disturbing
M Cozzolino et al.
The International journal of artificial organs, 30(4), 293-300 (2007-05-24)
Phosphate overload is a dramatic consequence in end-stage renal disease (ESRD) patients. Recent studies have well documented that abnormalities in mineral and bone metabolism in these patients are associated with increased cardiovascular morbidity and mortality. Elevated serum phosphate and calcium-phosphate
Gadolinium and lanthanum: a iatrogenic transmetallation?
Simona Brambilla et al.
Clinical biochemistry, 41(13), 1029-1033 (2008-06-19)
Declan de Freitas et al.
Seminars in dialysis, 20(4), 325-328 (2007-07-20)
Hyperphosphatemia is associated with increased morbidity and mortality in dialysis patients. Oral phosphate binders are necessary to control serum phosphate in patients eating a normal diet and undergoing peritoneal dialysis or thrice weekly hemodialysis. Until recently, none of the available
Marc E De Broe
Seminars in dialysis, 21(2), 142-144 (2008-02-06)
The recent recognition of gadolinium (Gd) administration as a cause of nephrogenic systemic fibrosis (NSF) is a growing concern. The similarity of Gd and lanthanum (La), based on their place in the table of Mendeljev, has led to assumptions that

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service