Skip to Content
Merck
All Photos(2)

Key Documents

GF08107602

Zirconium

foil, light tested, 25x25mm, thickness 0.075mm, annealed, 99.2%

Synonym(s):

Zirconium, ZR000240

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 081-076-02

resistivity

40 μΩ-cm, 20°C

L × W × thickness

25 mm × 25 mm × 0.075 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oluwatayo F Ikotun et al.
Future medicinal chemistry, 3(5), 599-621 (2011-04-30)
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted
Abnormal trace metals in man: zirconium.
H A Schroeder et al.
Journal of chronic diseases, 19(5), 573-586 (1966-05-01)
Daryoush Afzali et al.
Environmental monitoring and assessment, 186(6), 3523-3529 (2014-02-04)
In this study, a ternary Zr(IV) system with 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol (5-Br-PADAP) and fluoride was chosen on the basis of dispersive liquid-liquid microextraction method. Zirconium was extracted into the fine droplets of dichlorobenzene as extracting solvent. These drops dispersed as a cloud
Yugo Hatai
The international journal of esthetic dentistry, 9(2), 206-222 (2014-04-26)
In contemporary dentistry, we have a vast range of materials to choose from, and metal free restorations have become the premier materials for achieving the ultimate in both esthetics and durability. Metal-free restorations are utilized with more conservative preparations to
R Velázquez-Cayón et al.
Medicina oral, patologia oral y cirugia bucal, 17(2), e246-e250 (2011-12-07)
The increase of aesthetic demands, together with the successful outcome of current implants, has renewed interest in the search for new materials with enough mechanical properties and better aesthetic qualities than the materials customarily used in implanto-prosthetic rehabilitation. Among these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service