Skip to Content
Merck
All Photos(1)

Key Documents

F6206

Sigma-Aldrich

4-Fluorobenzenesulfonyl chloride

98%

Synonym(s):

4-Fluorobenzene-1-sulfonyl chloride, p-Fluorobenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2Cl
CAS Number:
Molecular Weight:
194.61
Beilstein:
2091633
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

95-96 °C/2 mmHg (lit.)

mp

29-31 °C (lit.)

SMILES string

Fc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C6H4ClFO2S/c7-11(9,10)6-3-1-5(8)2-4-6/h1-4H

InChI key

BFXHJFKKRGVUMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y A Chang et al.
Bioconjugate chemistry, 3(2), 200-202 (1992-03-01)
4-Fluorobenzenesulfonyl chloride (fosyl chloride), due to the strong electron-withdrawing property of its fluoride atom, is found to be an excellent activating agent for the covalent attachment of biologicals to a variety of solid supports (e.g. functionalized polystyrene microspheres, Sepharose beads
T H Liao et al.
Analytical biochemistry, 148(2), 365-375 (1985-08-01)
The reagent p-fluorobenzenesulfonyl chloride modifies the protein side chains of tyrosine, lysine, and histidine and the alpha-NH2 group. The p-fluorobenzenesulfonyl (Fbs-) group, identified by the 19F nuclear magnetic resonance signal, exhibits a different 19F chemical shift for each functional group
Umaprasana Ojha et al.
Langmuir : the ACS journal of surfaces and colloids, 25(11), 6319-6327 (2009-04-02)
Peptide surface modification of poly[(methyl methacrylate-co-hydroxyethyl methacrylate)-b-isobutylene-b-(methyl methacrylate-co-hydroxyethyl methacrylate)] P(MMA-co-HEMA)-b-PIB-b-P(MMA-co-HEMA) triblock copolymers with different HEMA/MMA ratios has been accomplished using an efficient synthetic procedure. The triblock copolymers were reacted with 4-fluorobenzenesulfonyl chloride (fosyl chloride) in pyridine to obtain the activated
Ning Li et al.
European journal of medicinal chemistry, 155, 531-544 (2018-06-18)
Ten novel symmetric 3,5-bis(arylidene)-4-piperidone derivatives (BAPs, 1-10) and fourteen dissymmetric BAPs (11-24) were synthesized and evaluated the cytotoxicity. All of the compounds have been screened for their anti-inflammatory activity characterized by evaluating their inhibitory effects on LPS-induced IL-6, TNF-α secretion.
Abdelaziz Houmam et al.
Physical chemistry chemical physics : PCCP, 14(1), 113-124 (2011-09-15)
Important aspects of the electrochemical reduction of a series of substituted arene sulfonyl chlorides are investigated. An interesting autocatalytic mechanism is encountered where the starting material is reduced both at the electrode and through homogeneous electron transfer from the resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service