Skip to Content
Merck
All Photos(2)

Key Documents

DWKBWP2000P

Azlon Polypropylene Wide Neck Round Bottles with Screw Cap

round translucent polypropylene bottle, capacity 2000 mL

Synonym(s):

BWP2000P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121818
NACRES:
NB.43
capacity:
2000 mL
material:
round translucent polypropylene bottle
wide-neck
packaging:
pkg of 5 ea

material

round translucent polypropylene bottle
wide-neck

packaging

pkg of 5 ea

manufacturer/tradename

Azlon BWP2000P

capacity

2000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Featuring a leakproof, linerless cap design, AZLON® wide neck polypropylene bottles are easy to fill and autoclavable at 121°C.

Application

Ideal for a wide range of liquid storage and sampling applications

Features and Benefits

Autoclavable at 121°C and suitable for food contact applications

Legal Information

AZLON is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service