DWK651502
Wheaton® Vacule® Lyophilization Ampule
Synonym(s):
651502
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
preconditioned (pre-scored)
packaging
pkg of 144 ea
manufacturer/tradename
WHEATON® 651502
capacity
1 mL
diam. × H
10 mm × 108 mm
Looking for similar products? Visit Product Comparison Guide
Application
Lyophilization
Features and Benefits
- Ideal for small volume lyophillization samples
- Pre-scored for easy opening</li><li>Heat sealed only
- Converted from Type I borosilicate glass tubing, ampules conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1.
Legal Information
Vacule is a registered trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service