Skip to Content
Merck
All Photos(2)

Key Documents

D94208

Sigma-Aldrich

Diethyl ethoxymethylenemalonate

99%

Synonym(s):

DEEMM, Ethoxymethylenemalonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=C(COOC2H5)2
CAS Number:
Molecular Weight:
216.23
Beilstein:
880058
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

279-281 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCO\C=C(\C(=O)OCC)C(=O)OCC

InChI

1S/C10H16O5/c1-4-13-7-8(9(11)14-5-2)10(12)15-6-3/h7H,4-6H2,1-3H3

InChI key

LTMHNWPUDSTBKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl ethoxymethylenemalonate can be used as a precursor to synthesize pyrimidinones, thiazolopyrimidines, thiazolopyrimidinones, triazolo [1, 5-a] pyrimidines, pyrimido[2,1-b]benzothiazoles and ethyl 3-amino-5-oxoisoxazolidine-4-carboxylate derivatives.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structure activity relationship investigation of triazolo [1, 5-a] pyrimidines as CB2 cannabinoid receptor inverse agonists.
Tabrizi M A, et al.
European Journal of Medicinal Chemistry, 113, 11-27 (2016)
Microwave Assisted Synthesis of Fused Thiazoles in Multicomponent System and Their in vitro Antitumor, Antioxidant, and Antimicrobial Activities.
El?Borai M A, et al.
Journal of Heterocyclic Chemistry, 54(2), 1031-1041 (2017)
Synthesis and in vitro antitumor activity of new series of benzothiazole and pyrimido [2, 1-b] benzothiazole derivatives.
Gabr M T, et al.
European Journal of Medicinal Chemistry, 85, 576-592 (2014)
An Efficient Cascade Synthesis of Ethyl 3-Amino-5-oxoisoxazolidine-4-carboxylate Derivatives.
Li R, et al.
Synthesis, 49(18), 4341-4349 (2017)
Synthesis of fused pyrimidinone and quinolone derivatives in an automated high-temperature and high-pressure flow reactor.
Tsoung J, et al.
The Journal of Organic Chemistry, 82(2), 1073-1084 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service