Skip to Content
Merck
All Photos(2)

Key Documents

D91152

Sigma-Aldrich

Diethyl 2-bromoethylphosphonate

97%

Synonym(s):

(2-Bromoethyl)phosphonic acid diethyl ester, NSC 119421, NSC 2672

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2P(O)(OCH2CH3)2
CAS Number:
Molecular Weight:
245.05
Beilstein:
1766077
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.461 (lit.)

bp

75 °C/1 mmHg (lit.)

density

1.348 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CCBr)OCC

InChI

1S/C6H14BrO3P/c1-3-9-11(8,6-5-7)10-4-2/h3-6H2,1-2H3

InChI key

PINITSMLVXAASM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Radical coupling
  • Synthesis of organosoluble zirconium phosphonate nanocomposites
  • Click-chemistry
  • Asymmetric epoxidation of unfunctionalized olefins
  • Negishi alkyl-aryl cross-coupling
  • Synthesis of hydrolytically stable phosphonic acids for use as dental adhesives

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service