D38308
2,4′-Dibromoacetophenone
>98%
Synonym(s):
4′-Bromophenacyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
BrC6H4COCH2Br
CAS Number:
Molecular Weight:
277.94
Beilstein:
607604
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
>98%
form
crystals
mp
108-110 °C (lit.)
SMILES string
BrCC(=O)c1ccc(Br)cc1
InChI
1S/C8H6Br2O/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2
InChI key
FKJSFKCZZIXQIP-UHFFFAOYSA-N
Gene Information
human ... PLA2G2A(5320)
rat ... Pla2g2a(29692)
Looking for similar products? Visit Product Comparison Guide
Application
Undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. Also useful in the esterification of carboxylic acids.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrahedron, 49, 9735-9735 (1993)
Synthetic Communications, 23, 271-271 (1993)
Tetrahedron Letters, 34, 4547-4547 (1993)
Angelo J Magro et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 12), 1670-1677 (2005-11-23)
The crystal structure of an acidic phospholipase A(2) isolated from Bothrops jararacussu venom (BthA-I) chemically modified with p-bromophenacyl bromide (BPB) has been determined at 1.85 Angstroms resolution. The catalytic, platelet-aggregation inhibition, anticoagulant and hypotensive activities of BthA-I are abolished by
Wen-Hsin Liu et al.
Toxicology letters, 185(2), 102-109 (2009-01-03)
In view of the controversial role of catalytic activity on the cytotoxicity of phospholipase A(2) (PLA(2)), the present study is conducted to explore whether PLA(2) induces apoptotic process of human leukemia U937 cells through catalytic activity-independent pathway. Modification of His-48
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service