Skip to Content
Merck
All Photos(1)

Key Documents

D186252

Sigma-Aldrich

N,N-Dimethylsulfamoyl chloride

99%

Synonym(s):

Dimethylamidosulfonyl chloride, Dimethylaminosulfonyl chloride, Dimethylsulphamoyl chloride, N,N-Dimethylamidosulfonyl chloride, N,N-Dimethylaminosulfonyl chloride, N,N-Dimethylsulfamoyl chloride, N,N-Dimethylsulfamyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NSO2Cl
CAS Number:
Molecular Weight:
143.59
Beilstein:
741979
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

114 °C/75 mmHg (lit.)

density

1.337 g/mL at 25 °C (lit.)

SMILES string

CN(C)S(Cl)(=O)=O

InChI

1S/C2H6ClNO2S/c1-4(2)7(3,5)6/h1-2H3

InChI key

JFCHSQDLLFJHOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Applications of N,N -dimethylsulfamoyl chloride are:
  • It is widely used in the synthesis of a medicinally important class of compounds, sulfonamides.
  • It is used in the synthesis of aminotetralin derived sulfamides as anticancer agents and acetylcholinesterase inhibitors.
  • It can also be used in the esterification/amidation between carboxylic acids and equimolar amounts of alcohols/amines, as in the synthesis of coumaperine.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel and efficient method for esterification, amidation between carboxylic acids and equimolar amounts of alcohols, and amines utilizing Me2NSO2Cl and N, N-dimethylamines; its application to the synthesis of coumaperine, a natural chemopreventive dieneamide.
Wakasugi K, et al.
Tetrahedron, 59(28), 5337-5345 (2003)
Acetylcholinesterase and carbonic anhydrase inhibitory properties of novel urea and sulfamide derivatives incorporating dopaminergic 2-aminotetralin scaffolds.
Ozgeris B, et al.
Bioorganic & Medicinal Chemistry, 24(10), 2318-2329 (2016)
Catalytic arylation of sulfamoyl chlorides: A practical synthesis of sulfonamides.
Frost CG, et al.
Synlett, 2002(11), 1928-1930 (2002)
Synthesis and Anticancer Activity of Novel Ureas and Sulfamides Incorporating 1-Aminotetralins.
Ozgeris B, et al.
Archives of Medical Research (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service