D124001
3,5-Diiodosalicylic acid
99%
Synonym(s):
2-Hydroxy-3,5-diiodobenzoic acid, 3,5-Diiodo-2-hydroxybenzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
I2C6H2(OH)CO2H
CAS Number:
Molecular Weight:
389.91
Beilstein:
2615358
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
mp
220-230 °C (dec.) (lit.)
SMILES string
OC(=O)c1cc(I)cc(I)c1O
InChI
1S/C7H4I2O3/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,10H,(H,11,12)
InChI key
DHZVWQPHNWDCFS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Z Avramova et al.
Plant molecular biology, 22(6), 1135-1143 (1993-09-01)
Nuclear matrices were isolated from maize leaves by the two conventional methods usually employed for the preparation of the corresponding structures of animal origin. It is demonstrated that functionally competent matrices, recognizing and specifically binding the MAR-containing DNA of the
J Suominen et al.
Archives of andrology, 15(2-3), 109-116 (1985-01-01)
Various methods of solubilization of human sperm antigens were compared by testing the sperm extracts against rabbit antihuman spermatozoa immune serums with double immunodiffusion and rocket immunoelectrophoresis. Marked differences were found in the protein contents and the SDS-PAGE protein patterns
Tamás Kovács et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 73(10), 965-970 (2008-08-02)
Rapid flow cytometric measurement of the frequency of aneuploid human sperms is in increasing demand but development of an exploitable method is hindered by difficulties of stoichiometric staining of sperm DNA. An aggressive decondensation protocol is needed after which cell
A B Czuppon et al.
Biochemistry international, 11(1), 21-34 (1985-07-01)
Five glycoproteinmolecules with the molecular masses of 17 000; 38 000; 42 000; 50 000 and 67 000 were purified by high performance liquid chromatography following solubilization of isolated porcine zonae pellucidae by treatment with lithium-3,5-diiodosalicylate. The N-terminal amino acid
R K Naz et al.
Archives of andrology, 29(2), 117-126 (1992-09-01)
Sperm samples were collected from male partners of fertile and infertile couples having unexplained and male factor infertility and analyzed for morphological abnormalities. After a staining procedure, sperm samples of these infertile men showed a variety of abnormalities. The sperm
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service