D109401
2,4-Dihydroxybenzoic acid
97%
Synonym(s):
β-Resorcylic acid
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1946213
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
208-211 °C (dec.) (lit.)
SMILES string
OC(=O)c1ccc(O)cc1O
InChI
1S/C7H6O4/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,8-9H,(H,10,11)
InChI key
UIAFKZKHHVMJGS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2,4-Dihydroxybenzoic acid (DHBA) can be used as a starting material for the synthesis of resorcinol-based N-benzyl benzamide derivatives , hydroxylated and prenylated xanthones , and mycophenolic acid.
It can be also used as a reactant in the synthesis of DHBARF polymer aerogels by reacting with formadehyde (F) and resorcinol (R).
It can be also used as a reactant in the synthesis of DHBARF polymer aerogels by reacting with formadehyde (F) and resorcinol (R).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Potential of 2,4-Dihydroxybenzoic Acid as an Oviposition Stimulant for Mass-Reared Ladybird Beetles.
E W Riddick et al.
Journal of insect science (Online), 19(2) (2019-03-02)
The discovery of inexpensive, readily available bioflavonoids, and their degradation products that boost the reproductive potential of mass-reared predators is the overarching goal of this research. We tested the hypothesis that 2,4-dihydroxybenzoic acid (DHBA), an inexpensive degradation product of morin
The synthesis of mycophenolic acid from 2, 4-dihydroxybenzoic acid
Patterson JW
The Journal of Organic Chemistry, 60(14), 4542-4548 (1995)
Synthesis, SAR and biological evaluation of natural and non-natural hydroxylated and prenylated xanthones as antitumor agents
Zhang X, et al.
Medicinal Chemistry, 8(6), 1012-1025 (2012)
Design, synthesis, and biological evaluation of a series of resorcinol-based N-benzyl benzamide derivatives as potent Hsp90 inhibitors
Park SY, et al.
European Journal of Medicinal Chemistry, 143, 390-401 (2018)
Jafar Ebrahimifar et al.
Insects, 11(9) (2020-09-05)
The ladybird beetle Stethorus gilvifrons (Mulsant) is an important natural enemy of tetranychid mites and functions as a biological control of these plant pests. The development, survival and reproduction of S. gilvifrons were studied when fed on three artificial diets.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service