Skip to Content
Merck
All Photos(2)

Key Documents

D0387

Sigma-Aldrich

6,7-Dimethyl-5,6,7,8-tetrahydropterine hydrochloride

≥95%

Synonym(s):

2-Amino-6,7-dimethyl-4-hydroxy-5,6,7,8-tetrahydropteridine, DMPH4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13N5O · HCl
CAS Number:
Molecular Weight:
231.68
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

Cl[H].CC1Nc2nc(N)nc(O)c2NC1C

InChI

1S/C8H13N5O.ClH/c1-3-4(2)11-6-5(10-3)7(14)13-8(9)12-6;/h3-4,10H,1-2H3,(H4,9,11,12,13,14);1H

InChI key

GIHYTRGUZVYCQX-UHFFFAOYSA-N

Biochem/physiol Actions

Synthetic reduced pterin cofactor for nitric oxide synthetase, and for phenylalanine, tyrosine, and tryptophan hydroxylases; less active than the natural cofactor, tetrahydrobiopterin (BH4).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Koizumi et al.
The Journal of antibiotics, 35(4), 458-462 (1982-04-01)
Phenylalanine hydroxylase was shown to be inhibited by oudenone and its derivatives in vitro. At a concentration of 2.3 x 10(-3) M, oudenone inhibited phenylalanine hydroxylase by 50%, and some of the oudenone derivatives showed more potent inhibition. The kinetic
C Unger et al.
Cancer research, 45(2), 616-618 (1985-02-01)
The O-alkyl cleavage enzyme is important for the metabolism of cytotoxic alkyl-lysophospholipids. We have developed a simple new method for the determination of the enzyme activity which is based on the formation of water-soluble phosphate during the enzyme reaction. This
H Nakata et al.
Journal of biochemistry, 90(2), 567-569 (1981-08-01)
A simple and rapid method for isolating tryptophan 5-monooxygenase [L-tryptophan, tetrahydropteridine:oxygen oxidoreductase (5-hydroxylating), EC 1.14.16.4] was reported. The method involves adsorption on calcium phosphate gel and affinity chromatography on agarose coupled with dimethyltetrahydropteridine. Tryptophan 5-monooxygenase was purified 1,100-fold from a
M A Parniak et al.
The Journal of biological chemistry, 263(3), 1223-1230 (1988-01-25)
The pH optimum of rat liver phenylalanine hydroxylase is dependent on the structure of the cofactor employed and on the state of activation of the enzyme. The tetrahydrobiopterin-dependent activity of native phenylalanine hydroxylase has a pH optimum of about 8.5.
I G Jennings et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(13), 5734-5738 (1991-07-01)
A monoclonal anti-idiotype antibody, NS7, previously shown to mimic the binding of the pterin cofactor of phenylalanine hydroxylase (phenylalanine 4-monooxygenase, EC 1.14.16.1) has been used to localize the cofactor binding site within the phenylalanine hydroxylase catalytic domain to a 27-amino-acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service