CLS6404250
Pyrex® Squibb separatory funnel, PE stopper, PTFE stopcock
capacity 250 mL
Synonym(s):
Corning® 6404
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
Joint
joint stopper22
manufacturer/tradename
Corning 6404-250
capacity
250 mL
size
stopcock size
4
General description
PYREX® pear-shaped glass separatory funnels contain stoppers and polytetrafluorethylene (PTFE) stopcocks. The closed bottom stopper made of linear high density polyethylene to conform to standard taper stopper dimensions. The PTFE plug is designed to fit into the micro-finish barrel and it also minimizes freezing problems and grease contamination.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)