Skip to Content
Merck
All Photos(1)

Key Documents

CDS018120

Sigma-Aldrich

6-hydroxy-2-naphthaldehyde

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Oc1ccc2cc(C=O)ccc2c1

InChI

1S/C11H8O2/c12-7-8-1-2-10-6-11(13)4-3-9(10)5-8/h1-7,13H

InChI key

PRYNJOJHKYNLIS-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bo Lin et al.
The Analyst, 143(20), 5054-5060 (2018-09-22)
A mitochondrial-targeted pH fluorescent probe 4-(2-(6-hydroxynaphthalen-2-yl)vinyl)-1-methylpyridin-1-ium was facilely synthesized via the carbon-carbon double bond bridging of 6-hydroxy-2-naphthaldehyde and 1,4-dimethylpyridinium iodide salt. The probe exhibited remarkable pH-dependent behavior in the linear range of 7.60-10.00, with a pKa value of 8.85 ±
Yongfei Wang et al.
Organic & biomolecular chemistry, 15(13), 2734-2739 (2017-01-18)
Although sulfur dioxide (SO
Hao Wang et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(11), 1297-1302 (2018-07-31)
A new highly selective and visible colorimetric fluorescent probe (probe 1) was developed to detect hydrazine (N2H4) concentration in real water samples. As different concentrations of N2H4 were added, the color of the probe solution was graded gradually from colorless
Zejun Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 230, 118073-118073 (2020-01-25)
Hypochlorous acid (HOCl) and peroxynitrite (ONOO-) are two of the most important reactive species and associated with various diseases in various physiological and pathological processes. Nonetheless, many of their roles are still vague due to the shortage of methods for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service