Skip to Content
Merck
All Photos(1)

Key Documents

CDS000572

Sigma-Aldrich

Sodium monododecyl phosphate

AldrichCPR

Synonym(s):

Sodium Monolauryl Phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25Na2O4P
Molecular Weight:
310.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

[Na+].[Na+].CCCCCCCCCCCCOP([O-])([O-])=O

InChI

1S/C12H27O4P.2Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;;/h2-12H2,1H3,(H2,13,14,15);;/q;2*+1/p-2

InChI key

YVIGPQSYEAOLAD-UHFFFAOYSA-L

Application

Sodium monododecyl phosphate is an ionic surfactant that can be used along with transglycosylated stevia (stevia-G) to enhance the solubility and bioavailability of poorly water-soluble drugs. It is also used to synthesize self-assembled electroactive nanofilms by complexation with polyaminobenzylamines for nanoarchitectonic applications.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mixed micelle system produced by interaction between transglycosylated stevia and an ionic surfactant improves dissolution profile of mefenamic acid
Fujimori M, et al.
Journal of Pharmaceutical Sciences, 106(4), 1117-1123 (2017)
Carla Atallah et al.
International journal of pharmaceutics, 589, 119721-119721 (2020-08-08)
Despite the high aqueous solubility of cysteamine, its unpleasant organoleptic properties, hygroscopicity, instability in solutions, and poor pharmacokinetic profile are the main drawbacks that limit its use for medical and cosmetic purposes. In this study, cysteamine-loaded liposomes were prepared using
John Oludele Olanlokun et al.
Drug and chemical toxicology, 44(1), 47-57 (2018-11-30)
Malaria is a parasitic disease that has defied many treatment plans. This study was carried out to investigate the host mitochondrial response to malarial infection and selected antimalarial chemotherapy using murine models. The effects of artesunate (ART) and proguanil (PRG)
Amir Jalali et al.
Journal of venom research, 3, 1-6 (2012-07-25)
Hemiscorpius lepturus (H. lepturus) is one of the most dangerous scorpions in Iran. Intramuscular administration (IM) of available Razi antivenom to H. lepturus venom is used by many of Iranian clinicians. The purpose of the current study was to investigate
Electrochemical nanoarchitectonics through polyaminobenzylamine-dodecyl phosphate complexes: redox activity and mesoscopic organization in self-assembled nanofilms
Lorenzo A, et al.
Physical Chemistry Chemical Physics, 20(11), 7570-7578 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service