Skip to Content
Merck
All Photos(1)

Key Documents

C91492

Sigma-Aldrich

Cyanogen bromide

reagent grade, 97%

Synonym(s):

Bromine cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCN
CAS Number:
Molecular Weight:
105.92
Beilstein:
1697296
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

vapor density

3.65 (vs air)

vapor pressure

100 mmHg ( 22.6 °C)

Assay

97%

form

crystals

bp

61-62 °C (lit.)

mp

50-53 °C (lit.)

storage temp.

2-8°C

SMILES string

BrC#N

InChI

1S/CBrN/c2-1-3

InChI key

ATDGTVJJHBUTRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyanogen bromide (CNBr) is a halogen halide. Its utility as a condensing agent during chemical ligation of oligodeoxyribonucleotides has been reported. Its threshold photoelectron (TPE) spectrum has been recorded under high resolution. The cryopolymerization of CNBr on irradiation has been investigated.

Application

Cyanogen bromide may be used:
  • To activate polysaccharides for coupling with erythrocytes.
  • As an oxidizing agent in urine thaimine assay.
  • In the preparation of 2-amino-5-(2′-thienyl)-1,3,4-oxadiazole.
For peptide cleavage and use in protein immobilization procedures. Reaction with C60Ph5Cl produces a novel phenylated isoquinolino[3′,4′:1,2][60]fullerene.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>149.0 °F - closed cup

Flash Point(C)

> 65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Org. Mass Spectrom., 28, 1395-1395 (1993)
Applied Biochemistry and Biotechnology, 44, 135-135 (1994)
Coupling of polysaccharides to erythrocytes by cyanogen bromide.
A W Richter et al.
Scandinavian journal of immunology, 3(3), 365-368 (1974-01-01)
Oxadiazole: a biologically important heterocycle.
Somani RR and Shirodkar PY.
Der Pharma Chemica, 1(1), 130-140 (2009)
Low-temperature radiation polymerization of cyanogen bromide.
Kichigina GA, et al.
Mendeleev Communications, 8(4), 159-160 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service