BR781300
BRAND® PCR tubes
capacity 0.2 mL, colorless tube, individual, attached domed cap
Synonym(s):
PCR, PCR Tubes, Tubes
About This Item
Recommended Products
material
colorless cap
colorless tube
conical bottom
domed cap
polypropylene cap
polypropylene tube
description
thin-walled
sterility
non-sterile
feature
graduations: no
individual, attached domed cap
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 781300
technique(s)
PCR: suitable
capacity
0.2 mL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Thin walls for rapid heat transfer
- Tightly closing covers protect samples against evaporation
- Colors available for easy identification of samples
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service