BR779012
BRAND® centrifuge tube, rimless
volume 15 mL, conical bottom, graduated: no
Synonym(s):
Centrifuge tubes, Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
AR-Glass
conical bottom
description
RCF up to 3500
sterility
non-sterile
feature
graduated: no
packaging
pack of 100
manufacturer/tradename
BRAND 779012
volume
15 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
TheAR-Glas® centrifuge tube offers a capacity of approximately 15 ml and features a sturdy wall thickness of 1.0 mm. Engineered for high-performance applications, it withstands relative centrifugal forces (RCF) of up to 3500. This reliable tube is perfect for laboratory use, ensuring durability and precision during centrifugation processes. An essential addition to any scientific workspace.
Legal Information
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service