Key Documents
BR701374
BRAND® sealing film for microplates
self-adhesive, suitable for storage, vinyl (with pre-pierced zone)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
material
film (PP/PE-HD)
vinyl (with pre-pierced zone)
sterility
non-sterile
feature
self-adhesive
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701374
suitability
suitable for storage
General description
Due to the increasing number of analysis methods, the number of samples processed is growing steadily. This means that high-throughput analyses are gaining significance. In these automated processes, the films must not only protect the samples, but also comply with the mechanical requirements of automation.
Features and Benefits
  • Pierceability
  • Adhesive-free zones for contamination-free tasks
  • Tight seal for minimizing evaporation
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)