BR701352
BRAND® 96-well deep well plate, stackable
volume 1.1 mL, polystyrene
Synonym(s):
Deep-well Plates, Sample Storage
All Photos(1)
About This Item
material
polystyrene
description
alphanumeric identification system
sterility
non-sterile
feature
skirt
packaging
pack of 32 ea
manufacturer/tradename
BRAND 701352
size
96 well
volume
1.1 mL
working volume
1.1 mL
binding type
non-treated surface
General description
- BRAND® 96-well deep well plates are made of high purity polystyrene (PS).
- The ′U′ bottom ensures optimal sample mixture and collection.
- Designed for a range of applications including High-Throughput Screening (HTS), serial dilutions, reagent transfer, and sample storage down to -80 °C.
- Stackable for easy storage.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG