Skip to Content
Merck
All Photos(1)

Key Documents

BAH377740000

Stirring bar retriever

white PTFE (flexible), diam. × L 12.5 in. (53 mm) × 2.0625 in. , magnet

Synonym(s):

Stirring bar retriever

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43
Pricing and availability is not currently available.

material

white PTFE (flexible)

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H37774-0000

diam. × L

12.5 in. (53 mm) × 2.0625 in. , magnet

Looking for similar products? Visit Product Comparison Guide

General description

Flexible PTFE rod with a powerful Alnico PTFE-coated magnet for retrieving magnetic stirring bars from containers or non-magnetic sink drains and traps.

Features and Benefits

  • Flexible white rod is 33cm (13") long
  • Autoclavable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service