Skip to Content
Merck
All Photos(1)

Key Documents

B91005

Sigma-Aldrich

tert-Butyl carbazate

98%, for peptide synthesis

Synonym(s):

tert-Butoxycarbonyl hydrazide, Boc-hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHNH2
CAS Number:
Molecular Weight:
132.16
Beilstein:
1756967
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

tert-Butyl carbazate, 98%

Assay

98%

form

crystals

bp

63-65 °C/0.1 mmHg (lit.)

mp

39-42 °C (lit.)

application(s)

peptide synthesis

SMILES string

O=C(NN)OC(C)(C)C

InChI

1S/C5H12N2O2/c1-5(2,3)9-4(8)7-6/h6H2,1-3H3,(H,7,8)

InChI key

DKACXUFSLUYRFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a palladium-catalyzed cross-coupling with vinyl halides leading to N-Boc-N-alkenylhydrazines.[1]
Reagent used in solid phase peptide synthesis[2] and in α-amino aldehyde optical purity determinations.[3] Condenses with aldehydes to form hydrazones which are intermediates in the synthesis of HIV-1 protease inhibitors.[4]

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Letters, 34, 6599-6599 (1993)
Journal of the Chemical Society. Chemical Communications, 1052-1052 (1993)
José Barluenga et al.
Organic letters, 9(2), 275-278 (2007-01-16)
N-Boc-N-alkenylhydrazines, an almost unknown type of compounds, have been prepared with high to moderate yields via palladium-catalyzed cross-coupling between alkenyl halides and tert-butyl carbazate. The present methodology represents the first general way to access this highly functionalized and unusual type
Tetrahedron Letters, 34, 5425-5425 (1993)
Martina Bruna Violatto et al.
ACS nano, 13(4), 4410-4423 (2019-03-19)
Steroids are the standard therapy for autoimmune hepatitis (AIH) but the long-lasting administration is hampered by severe side effects. Methods to improve the tropism of the drug toward the liver are therefore required. Among them, conjugation to nanoparticles represents one

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service