Skip to Content
Merck
All Photos(2)

Key Documents

B81956

Sigma-Aldrich

4-Bromothiophenol

95%

Synonym(s):

4-Bromobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4SH
CAS Number:
Molecular Weight:
189.07
Beilstein:
507031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

crystals

bp

239 °C (lit.)

mp

73-76 °C (lit.)

SMILES string

Sc1ccc(Br)cc1

InChI

1S/C6H5BrS/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

FTBCOQFMQSTCQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aviva Levina et al.
Journal of the American Chemical Society, 132(25), 8720-8731 (2010-06-10)
The nature of the long-lived EPR-active Cr(V) species observed in cells and biological fluids exposed to carcinogenic Cr(VI) has been definitively assigned from detailed kinetic and spectroscopic analyses of a model reaction of Cr(VI) with p-bromobenzenethiol (RSH) in the presence
Homan Kang et al.
International journal of molecular sciences, 22(4) (2021-02-14)
We present a template-assisted method for synthesizing nanogap shell structures for biomolecular detections based on surface-enhanced Raman scattering. The interior nanogap-containing a silver shell structure, referred to as a silver nanogap shell (Ag NGS), was fabricated on silver nanoparticles (Ag
Myeong Geun Cha et al.
Nanoscale, 9(34), 12556-12564 (2017-08-19)
Multimodal imaging can provide complementary biomedical information which has huge potential in pre-clinical and clinical imaging and sensing. In this study, we introduce dual modal NIR silver bumpy nanoprobes for in vivo imaging and multiplexed detection of biomolecules by both
Hyung-Mo Kim et al.
Scientific reports, 8(1), 13938-13938 (2018-09-19)
In this study, we report on the fabrication of multilayered tri-functional magnetic-SERS-fluorescence nanoprobes (MF-SERS particles) containing clustered superparamagnetic Fe3O4 nanoparticles (NPs), silver NPs, and a fluorescent silica layer. The MF-SERS particles exhibited strong SERS signals from the silver NPs as
R Fliege et al.
Chemical research in toxicology, 13(5), 363-372 (2000-05-17)
The mycotoxin patulin (PAT) is believed to exert its cytotoxic and chromosome-damaging effects by forming covalent adducts with essential cellular thiols. Since the chemical structures of such adducts are unknown to date, we have studied the reaction of PAT and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service