Skip to Content
Merck
All Photos(3)

Key Documents

A29585

Sigma-Aldrich

Allyl bromide

ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer

Synonym(s):

3-Bromo-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Br
CAS Number:
Molecular Weight:
120.98
Beilstein:
605308
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.2 (vs air)

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

554 °F

contains

≤1000 ppm propylene oxide as stabilizer

expl. lim.

7.3 %

refractive index

n20/D 1.469 (lit.)

bp

70-71 °C (lit.)

mp

−119 °C (lit.)

density

1.398 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrCC=C

InChI

1S/C3H5Br/c1-2-3-4/h2H,1,3H2

InChI key

BHELZAPQIKSEDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyl bromide can be used as a reagent to prepare:
  • Allyl ketones by Barbier-type allylation of various nitriles in the presence of Lewis acid.
  • Homoallylic alcohols by reacting with aldehydes or ketones using zinc dust as a catalyst.
  • 1,5-hexadiene by reductive C-C bond coupling using silver hydride cluster.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase synthesis of [Ag4H]+ and its mediation of the C-C coupling of allyl bromide
Khairallah GN and O'Hair, Richard AJ
Angewandte Chemie (International Edition in English), 44(5), 728-731 (2005)
Facile and efficient synthesis of homoallylic alcohols using allyl bromide and commercial zinc dust
Ranu BC, et al.
Tetrahedron Letters, 36(27), 4885-4888 (1995)
Synthesis of allyl ketone via Lewis acid promoted Barbier-type reaction
Lee Adam S-Y and Lin L-S
Tetrahedron Letters, 41(45), 8803-8806 (2000)
Jakub Wręczycki et al.
Materials (Basel, Switzerland), 13(11) (2020-06-11)
The superior ability of thiiranes (episulfides) to undergo ring-opening polymerization (ROP) in the presence of anionic initiators allows the preparation of chemically stable polysulfide homopolymers. Incorporation of elemental sulfur (S8) by copolymerization below the floor temperature of S8 permits the
José C González-Gómez et al.
The Journal of organic chemistry, 75(18), 6308-6311 (2010-08-21)
The combination of an aldehyde, an allylic bromide, and tert-butanesulfinamide in the presence of indium metal and titanium tetraethoxide allows straightforward access to homoallylamine derivatives in high yields and stereoselectivities. Moreover, the synthetic utility of the enantioenriched homoallylamine derived from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service