Skip to Content
Merck
All Photos(2)

Key Documents

A1577

Sigma-Aldrich

Aluminum hydroxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)3 · xH2O
CAS Number:
Molecular Weight:
78.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

concentration

50-57.5% (Al2O3, EDTA titration)

application(s)

battery manufacturing

SMILES string

O.O[Al](O)O

InChI

1S/Al.4H2O/h;4*1H2/q+3;;;;/p-3

InChI key

SMYKVLBUSSNXMV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joung-Wook Seo et al.
PloS one, 8(2), e56447-e56447 (2013-03-02)
The World Health Organization reports that 235 million people are currently affected by asthma. This disease is associated with an imbalance of Th1 and Th2 cells, which results in the upregulation of cytokines that promote chronic inflammation of the respiratory
Takayuki Nagai et al.
Evidence-based complementary and alternative medicine : eCAM, 2011, 604196-604196 (2009-10-29)
Effects of a Kampo (Japanese herbal) medicine "shoseiryuto (SST, xiao-qing-long-tang in Chinese)", which has been used for the treatment of allergic bronchial asthma clinically, were examined on ovalbumin (OVA)-sensitized allergic airway inflammation model (i.e., bronchial asthma) in a mouse. When
Florian Wolschin et al.
Plant methods, 1(1), 9-9 (2005-11-08)
Protein phosphorylation is accepted as a major regulatory pathway in plants. More than 1000 protein kinases are predicted in the Arabidopsis proteome, however, only a few studies look systematically for in vivo protein phosphorylation sites. Owing to the low stoichiometry
Shuo-Yao Qu et al.
Scandinavian journal of immunology, 91(5), e12865-e12865 (2020-03-19)
Plasmacytoid dendritic cells (pDCs) regulate immunity and promote tolerance in asthma. Notch signalling is a highly conserved pathway that regulates the immune response; however, its role in pDC-mediated asthmatic airway inflammation is unclear. This study clarified the effects of Notch
Kazufumi Matsushita et al.
Journal of immunology (Baltimore, Md. : 1950), 193(12), 5791-5800 (2014-11-05)
Allergen-specific IgE is linked to asthma pathogenesis, but the underlying mechanisms of IgE production in response to allergen exposure are poorly understood. In this article, we show that B cell-intrinsic MyD88 is essential for IgE/IgG1 production evoked by ragweed pollen

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service